MilliporeSigma
  • Receptor selectivity of Met-enkephalin-Arg6-Phe7, an endogenous opioid peptide, in cerebral cortex of human and rat.
Receptor selectivity of Met-enkephalin-Arg6-Phe7, an endogenous opioid peptide, in cerebral cortex of human and rat.
Life sciences (2000-07-15)
K W Kim, R S Woo, C J Kim, Y P Cheong, J K Kim, J Kwun, K P Cho
ABSTRACT
This study was undertaken to examine the receptor selectivity of Met-enkephalin-Arg6-Phe7 (MERF) employing radioreceptor binding assays in human cerebral cortex membranes, and to elucidate the responsible receptors that mediate the regulatory action of MERF on high (20 mM) K+-stimulated release of [3H]norepinephrine ([3H]-NE) in rat cortex slices. Specific binding of [3H]MERF was inhibited by DAMGO, Tyr-D-Arg-Phe-Sar(TAPS), bremazocine and ethylketocyclazocine (EKC), but not by U69,593 (U69) and DPDPE. MERF showed high affinity for specific binding sites of [3H]DAMGO. However, MERF had little influence on the specific binding of [3H]DPDPE, [3H]U69 and [3H]diprenorphine ([3H]DIP) in the presence of 1 microM each of DAMGO, DPDPE and U69. In [3H]NE release experiments using rat cortex slices, DAMGO, MERF and EKC, in order of their potency, inhibited K+-stimulated release of [3H]NE. The inhibitory effects of MERF and DAMGO were more sensitive than that of EKC to antagonism by CTAP, nor-binaltorphimine (nor-BNI) and naloxone. These results suggested that MERF possesses high affinity for mu-receptors, but not for delta-, kappa1-, and very low affinity for kappa2-receptors in human cerebral cortex membranes. Also, the inhibitory effect of MERF on the K+-stimulated release of [3H]NE appears to be mediated by mu-receptors in rat cerebral cortex slices.