MilliporeSigma
  • High Endothelial Venules Accelerate Naive T Cell Recruitment by Tumor Necrosis Factor-Mediated R-Ras Upregulation.
High Endothelial Venules Accelerate Naive T Cell Recruitment by Tumor Necrosis Factor-Mediated R-Ras Upregulation.
The American journal of pathology (2020-11-08)
Junko Sawada, Carole Y Perrot, Linyuan Chen, Ashley E Fournier-Goss, Jeremiah Oyer, Alicja Copik, Masanobu Komatsu
ABSTRACT
Recruitment of naive T cells to lymph nodes is essential for the development of adaptive immunity. Upon pathogen infection, lymph nodes promptly increase the influx of naive T cells from the circulation in order to screen and prime the T cells. The precise contribution of the lymph node vasculature to the regulation of this process remains unclear. Here we show a role for the Ras GTPase, R-Ras, in the functional adaptation of high endothelial venules to increase naive T cell trafficking to the lymph nodes. R-Ras is transiently up-regulated in the endothelium of high endothelial venules by the inflammatory cytokine tumor necrosis factor (TNF) within 24 hours of pathogen inoculation. TNF induces R-Ras upregulation in endothelial cells via JNK and p38 mitogen-activated protein kinase but not NF-κB. Studies of T cell trafficking found that the loss of function of endothelial R-Ras impairs the rapid acceleration of naive T cell recruitment to the lymph nodes upon inflammation. This defect diminished the ability of naive OT-1 T cells to develop antitumor activity against ovalbumin-expressing melanoma. Proteomic analyses suggest that endothelial R-Ras facilitates TNF-dependent transendothelial migration (diapedesis) of naive T cells by modulating molecular assembly the at T cell-endothelial cell interface. These findings give new mechanistic insights into the functional adaptation of high endothelial venules to accelerate naive T cell recruitment to the lymph nodes.
Sigma-Aldrich
Anti-CDC42EP1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution