MilliporeSigma
  • Identification of two related pentapeptides from the brain with potent opiate agonist activity.
Identification of two related pentapeptides from the brain with potent opiate agonist activity.
Nature (1975-12-18)
J Hughes, T W Smith, H W Kosterlitz, L A Fothergill, B A Morgan, H R Morris
ABSTRACT
Enkephalin, a natural ligand for opiate receptors is composed of the pentapepides H-Tyr-Gly-Gly-Phe-Met-OH and H-Tyr-Gly-Gly-Phe-Leu-OH. The evidence is based on the determination of the amino acid sequence of natural enkephalin by the dansyl-Edman procedure and by mass spectrometry followed by synthesis and comparison of the natural and synthetic peptides.
Sigma-Aldrich
Leucine Enkephalin acetate salt hydrate, ≥95% (HPLC)
Pricing and availability is not currently available.