MilliporeSigma
  • SIRT1 and AMPK in regulating mammalian senescence: a critical review and a working model.
SIRT1 and AMPK in regulating mammalian senescence: a critical review and a working model.
FEBS letters (2010-12-07)
Yu Wang, Yan Liang, Paul M Vanhoutte
ABSTRACT
Ageing in mammals remains an unsolved mystery. Anti-ageing is a recurring topic in the history of scientific research. Lifespan extension evoked by Sir2 protein in lower organisms has attracted a large amount of interests in the last decade. This review summarizes recent evidence supporting the role of a Sir2 mammalian homologue, SIRT1 (Silent information regulator T1), in regulating ageing and cellular senescence. The various signaling networks responsible for the anti-ageing and anti-senescence activity of SIRT1 have been discussed. In particular, a counter-balancing model involving the cross-talks between SIRT1 and AMP-activated protein kinase (AMPK), another stress and energy sensor, is suggested for controlling the senescence program in mammalian cells.