MilliporeSigma
  • Identification of SPRED2 (sprouty-related protein with EVH1 domain 2) as a negative regulator of the hypothalamic-pituitary-adrenal axis.
Identification of SPRED2 (sprouty-related protein with EVH1 domain 2) as a negative regulator of the hypothalamic-pituitary-adrenal axis.
The Journal of biological chemistry (2011-01-05)
Melanie Ullrich, Karin Bundschu, Peter M Benz, Marco Abesser, Ruth Freudinger, Tobias Fischer, Julia Ullrich, Thomas Renné, Ulrich Walter, Kai Schuh
ABSTRACT
Sprouty-related proteins with EVH1 (enabled/vasodilator-stimulated phosphoprotein homology 1) domain (SPREDs) are inhibitors of MAPK signaling. To elucidate SPRED2 in vivo function, we characterized body homeostasis in SPRED2(-/-) mice. They showed a doubled daily water uptake, induced by elevated serum osmolality, originating from increased blood salt load. Accordingly, serum aldosterone was doubled, accompanied by augmented adrenal aldosterone synthase (AS) expression. Surprisingly, serum vasopressin (AVP) was unaltered, and, as evidenced by halved angiotensin II (Ang II) levels, the renin angiotensin system (RAS) was down-regulated. Adrenocorticotropic hormone (ACTH) was significantly elevated in SPRED2(-/-) mice, together with its secretagogue corticotropin-releasing hormone (CRH) and its downstream target corticosterone. ERK phosphorylation in brains was augmented, and hypothalamic CRH mRNA levels were elevated, both contributing to the increased CRH release. Our data were supported by CRH promoter reporter assays in hypothalamic mHypoE-44 cells, revealing a SPRED-dependent inhibition of Ets (ERK/E-twenty-six)-dependent transcription. Furthermore, SPRED suppressed CRH production in these cells. In conclusion, our study suggests that SPRED2 deficiency leads to an increased MAPK signaling, which results in an augmented CRH promoter activity. The subsequent CRH overproduction causes an up-regulation of downstream hypothalamic-pituitary-adrenal (HPA) hormone secretion. This constitutes a possible trigger for the observed compulsive grooming in SPRED2(-/-) mice and may, together with hyperplasia of aldosterone-producing cells, contribute to the hyperaldosteronism and homeostatic imbalances.