MilliporeSigma
  • Mitochondrial Damage-associated Molecular Patterns as Potential Biomarkers in DCD Heart Transplantation: Lessons From Myocardial Infarction and Cardiac Arrest.
Mitochondrial Damage-associated Molecular Patterns as Potential Biomarkers in DCD Heart Transplantation: Lessons From Myocardial Infarction and Cardiac Arrest.
Transplantation direct (2021-12-23)
Sarah L Longnus, Nina Rutishauser, Mark N Gillespie, Tobias Reichlin, Thierry P Carrel, Maria N Sanz
ABSTRACT
Heart transplantation with donation after circulatory death (DCD) has become a real option to increase graft availability. However, given that DCD organs are exposed to the potentially damaging conditions of warm ischemia before procurement, new strategies for graft evaluation are of particular value for the safe expansion of DCD heart transplantation. Mitochondria-related parameters are very attractive as biomarkers because of their intimate association with cardiac ischemia-reperfusion injury. In this context, a group of mitochondrial components, called mitochondrial damage-associated molecular patterns (mtDAMPs), released by stressed cells, holds great promise. mtDAMPs may be released at different stages of DCD cardiac donation and may act as indicators of graft quality. Because of the lack of information available for DCD grafts, we consider that relevant information can be obtained from other acute cardiac ischemic conditions. Thus, we conducted a systematic review of original research articles in which mtDAMP levels were assessed in the circulation of patients with acute myocardial infarction and cardiac arrest. We conclude that 4 mtDAMPs, ATP, cytochrome c, mitochondrial DNA, and succinate, are rapidly released into the circulation after the onset of ischemia, and their concentrations increase with reperfusion. Importantly, circulating levels of mtDAMPs correlate with cardiac damage and may be used as prognostic markers for patient survival in these conditions. Taken together, these findings support the concept that mtDAMPs may be of use as biomarkers to assess the transplant suitability of procured DCD hearts, and ultimately aid in facilitating the safe, widespread adoption of DCD heart transplantation.