MilliporeSigma
  • Genistein as a dietary supplement; formulation, analysis and pharmacokinetics study.
Genistein as a dietary supplement; formulation, analysis and pharmacokinetics study.
PloS one (2021-04-28)
Alexandra Mamagkaki, Ioannis Bouris, Panagiotis Parsonidis, Ioanna Vlachou, Maria Gougousi, Ioannis Papasotiriou
ABSTRACT
The objective of this study is to improve and optimize the formulation of Genistein in capsules in order to result in a better pharmacokinetic profile comparing to existing commercial products. In order to do this, five different formulations of Genistein capsules were developed and examined by reviewing their disintegration and dissolution properties. Furthermore, flowability of the powder along with potent incompatibilities between Genistein and its excipients were monitored through their thermal properties. The final formulation of Genistein was quantified using HPLC analysis and then its stability was evaluated thoroughly in real time and accelerated conditions. Finally, with the target to have a product with actual results, in vitro and in vivo studies were conducted. The final product proved to have better results in disintegration and dissolution. Moreover, R.G.C.C.'s capsules exhibited enhanced action in human cell lines as well as impressive pharmacokinetic results in animal models. The in vitro results showed an advantage of the R.G.C.C. product compared to the commercial one, whereas its maximum concertation in vivo was determined 34% higher than the commercial one.
Product Description
Supelco
Genistein, analytical standard
Pricing and availability is not currently available.