MilliporeSigma
VEGF receptor signalling - in control of vascular function.
Nature reviews. Molecular cell biology (2006-04-25)
Anna-Karin Olsson, Anna Dimberg, Johan Kreuger, Lena Claesson-Welsh
ABSTRACT
Vascular endothelial growth-factor receptors (VEGFRs) regulate the cardiovascular system. VEGFR1 is required for the recruitment of haematopoietic precursors and migration of monocytes and macrophages, whereas VEGFR2 and VEGFR3 are essential for the functions of vascular endothelial and lymphendothelial cells, respectively. Recent insights have shed light onto VEGFR signal transduction and the interplay between different VEGFRs and VEGF co-receptors in development, adult physiology and disease.