MilliporeSigma
  • Tumor-induced MDSC act via remote control to inhibit L-selectin-dependent adaptive immunity in lymph nodes.
Tumor-induced MDSC act via remote control to inhibit L-selectin-dependent adaptive immunity in lymph nodes.
eLife (2016-12-09)
Amy W Ku, Jason B Muhitch, Colin A Powers, Michael Diehl, Minhyung Kim, Daniel T Fisher, Anand P Sharda, Virginia K Clements, Kieran O'Loughlin, Hans Minderman, Michelle N Messmer, Jing Ma, Joseph J Skitzki, Douglas A Steeber, Bruce Walcheck, Suzanne Ostrand-Rosenberg, Scott I Abrams, Sharon S Evans
ABSTRACT
Myeloid-derived suppressor cells (MDSC) contribute to an immunosuppressive network that drives cancer escape by disabling T cell adaptive immunity. The prevailing view is that MDSC-mediated immunosuppression is restricted to tissues where MDSC co-mingle with T cells. Here we show that splenic or, unexpectedly, blood-borne MDSC execute far-reaching immune suppression by reducing expression of the L-selectin lymph node (LN) homing receptor on naïve T and B cells. MDSC-induced L-selectin loss occurs through a contact-dependent, post-transcriptional mechanism that is independent of the major L-selectin sheddase, ADAM17, but results in significant elevation of circulating L-selectin in tumor-bearing mice. Even moderate deficits in L-selectin expression disrupt T cell trafficking to distant LN. Furthermore, T cells preconditioned by MDSC have diminished responses to subsequent antigen exposure, which in conjunction with reduced trafficking, severely restricts antigen-driven expansion in widely-dispersed LN. These results establish novel mechanisms for MDSC-mediated immunosuppression that have unanticipated implications for systemic cancer immunity.
Sigma-Aldrich
PF-05480090, ≥98% (HPLC)
Pricing and availability is not currently available.