MilliporeSigma
  • Myoglobin on silica: a case study of the impact of adsorption on protein structure and dynamics.
Myoglobin on silica: a case study of the impact of adsorption on protein structure and dynamics.
Langmuir : the ACS journal of surfaces and colloids (2013-10-03)
Stéphanie Devineau, Jean-Marc Zanotti, Camille Loupiac, Loussiné Zargarian, Fabrice Neiers, Serge Pin, Jean Philippe Renault
ABSTRACT
If protein structure and function changes upon adsorption are well documented, modification of adsorbed protein dynamics remains a blind spot, despite its importance in biological processes. The adsorption of metmyoglobin on a silica surface was studied by isotherm measurements, microcalorimetry, circular dichroïsm, and UV-visible spectroscopy to determine the thermodynamic parameters of protein adsorption and consequent structure modifications. The mean square displacement and the vibrational densities of states of the adsorbed protein were measured by elastic and inelastic neutron scattering experiments. A decrease of protein flexibility and depletion in low frequency modes of myoglobin after adsorption on silica was observed. Our results suggest that the structure loss itself is not the entropic driving force of adsorption.
Sigma-Aldrich
Sodium phosphate dibasic dodecahydrate, BioXtra, ≥99.0% (T)