MilliporeSigma
  • A tetra-substituted chrysene: orientation of multiple electrophilic substitution and use of a tetra-substituted chrysene as a blue emitter for OLEDs.
A tetra-substituted chrysene: orientation of multiple electrophilic substitution and use of a tetra-substituted chrysene as a blue emitter for OLEDs.
Chemical communications (Cambridge, England) (2008-05-14)
Alex S Ionkin, William J Marshall, Brian M Fish, Lois M Bryman, Ying Wang
ABSTRACT
The first tetra-substituted non-fused chrysene, 3,6,9,12-tetrakis(4-tert-butylphenyl)chrysene with blue electroluminescence at 450 nm, and with a radiance of 500 cd m(-2), was synthesized by a two-step procedure: direct bromination of chrysene in trimethyl phosphate, followed by palladium-catalyzed cross-coupling of tetrabromochrysene and tert-butylphenylboronic acid .
Sigma-Aldrich
Chrysene