MilliporeSigma
  • Effect of the Addition Frequency of 5-Azacytidine in Both Micro- and Macroscale Cultures.
Effect of the Addition Frequency of 5-Azacytidine in Both Micro- and Macroscale Cultures.
Cellular and molecular bioengineering (2021-02-27)
Sandeep Kadekar, Laurent Barbe, Martin Stoddart, Oommen P Varghese, Maria Tenje, Gemma Mestres
ABSTRACT
Human mesenchymal stem cells (hMSCs) have a great clinical potential for tissue regeneration purposes due to its multilineage capability. Previous studies have reported that a single addition of 5-azacytidine (5-AzaC) causes the differentiation of hMSCs towards a myocardial lineage. The aim of this work was to evaluate the effect of 5-AzaC addition frequency on hMSCs priming (i.e., indicating an early genetic differentiation) using two culture environments. hMSCs were supplemented with 5-AzaC while cultured in well plates and in microfluidic chips. The impact of 5-AzaC concentration (10 and 20 μM) and addition frequency (once, daily or continuously), as well as of culture period (2 or 5 days) on the genetic upregulation of PPARγ (adipocytes), PAX3 (myoblasts), SOX9 (chondrocytes) and RUNX2 (osteoblasts) was evaluated. Daily delivering 5-AzaC caused a higher upregulation of PPARγ, SOX9 and RUNX2 in comparison to a single dose delivery, both under static well plates and dynamic microfluidic cultures. A particularly high gene expression of PPARγ (tenfold-change) could indicate priming of hMSCs towards adipocytes. Both macro- and microscale cultures provided results with similar trends, where addition frequency of 5-AzaC was a crucial factor to upregulate several genes. Microfluidics technology was proven to be a suitable platform for the continuous delivery of a drug and could be used for screening purposes in tissue engineering research.
Sigma-Aldrich
5-Azacytidine, ≥98% (HPLC)
Sigma-Aldrich
Gelatin from porcine skin, Type A, lyophilized powder, γ-irradiated, BioXtra, suitable for cell culture