MilliporeSigma
  • Active [3H]-dopamine uptake by human lymphocytes: correlates with serotonin transporter activity.
Active [3H]-dopamine uptake by human lymphocytes: correlates with serotonin transporter activity.
Pharmacology (1994-05-01)
B A Faraj, Z L Olkowski, R T Jackson
ABSTRACT
The main objective of the present investigation was to determine whether the uptake of [3H]-dopamine in human lymphocytes is mediated through a serotonin transporter. This was examined by studying the effects of various monoamine uptake inhibitors on the uptake of [3H]-dopamine in human lymphocytes. Among the compounds tested, indatraline, imipramine and fluoxetine, selective inhibitors of neuronal serotonin transporter, were the most potent inhibitors of [3H]-dopamine uptake in lymphocytes. The 50% inhibiting concentration (IC50) for these inhibitors was in the range of 3.5-17 nmol/l. Bupropion, GBR 12909, nomifensine and xylamine, selective inhibitors of dopamine and norepinephrine transporters, had low affinity for the dopamine uptake system in human lymphocytes with IC50 values ranging between 1,000 and 40,000 nmol/l. These findings provide supportive evidence for the participation of a serotonin transporter in the uptake of [3H]-dopamine in human lymphocytes. The existence of a high affinity transport system for dopamine and serotonin in human lymphocytes may serve as a readily accessible model to detect changes in the neuronal uptake of dopamine and serotonin in addictive and psychiatric disorders.