MilliporeSigma
  • NLRC4 inflammasome-dependent cell death occurs by a complementary series of three death pathways and determines lethality in mice.
NLRC4 inflammasome-dependent cell death occurs by a complementary series of three death pathways and determines lethality in mice.
Science advances (2021-10-23)
Peipei Zhang, Yifei Liu, Lichen Hu, Kai Huang, Mao Hong, Yuze Wang, Xinrui Fan, Richard J Ulevitch, Jiahuai Han
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Lipopolysaccharides from Escherichia coli K-235, purified by gel-filtration chromatography