MilliporeSigma
Basic methods for fission yeast.
Yeast (Chichester, England) (2006-02-25)
Susan L Forsburg, Nicholas Rhind
ABSTRACT
The fission yeast Schizosaccharomyces pombe is a popular model system, and has been particularly influential in studies of the cell cycle and chromosome dynamics. Despite its differences from Saccharomyces cerevisiae, the tools and methods for fission yeast are conceptually similar to those used in budding yeast. Here, we present basic methods sufficient for a beginner in this system to carry out most required manipulations for genetic analysis or molecular biology.