MilliporeSigma
CD36 is important for adipocyte recruitment and affects lipolysis.
Obesity (Silver Spring, Md.) (2013-03-21)
Irene O C M Vroegrijk, Jan Bert van Klinken, Janna A van Diepen, Sjoerd A A van den Berg, Maria Febbraio, Laura K M Steinbusch, Jan F C Glatz, Louis M Havekes, Peter J Voshol, Patrick C N Rensen, Ko Willems van Dijk, Vanessa van Harmelen
ABSTRACT
The scavenger receptor CD36 facilitates the cellular uptake of long-chain fatty acids. As CD36-deficiency attenuates the development of high fat diet (HFD)-induced obesity, the role of CD36-deficiency in preadipocyte recruitment and adipocyte function was set out to characterize. Fat cell size and number were determined in gonadal, visceral, and subcutaneous adipose tissue of CD36(-/-) and WT mice after 6 weeks on HFD. Basal lipolysis and insulin-inhibited lipolysis were investigated in gonadal adipose tissue. CD36(-/-) mice showed a reduction in adipocyte size in all fat pads. Gonadal adipose tissue also showed a lower total number of adipocytes because of a lower number of very small adipocytes (diameter <50 μm). This was accompanied by an increased pool of preadipocytes, which suggests that CD36-deficiency reduces the capacity of preadipocytes to become adipocytes. Regarding lipolysis, in adipose tissue from CD36(-/-) mice, cAMP levels were increased and both basal and 8-bromo-cAMP stimulated lipolysis were higher. However, insulin-mediated inhibition of lipolysis was more potent in CD36(-/-) mice. These results indicate that during fat depot expansion, CD36-deficiency negatively affects preadipocyte recruitment and that in mature adipocytes, CD36-deficiency is associated with increased basal lipolysis and insulin responsiveness.