MilliporeSigma
  • Immunohistochemical evaluation for P53 and VEGF (Vascular Endothelial Growth Factor) is not prognostic for long term survival in end stage esophageal adenocarcinoma.
Immunohistochemical evaluation for P53 and VEGF (Vascular Endothelial Growth Factor) is not prognostic for long term survival in end stage esophageal adenocarcinoma.
Revista do Colegio Brasileiro de Cirurgioes (2010-01-16)
Leandro Totti Cavazzola, André Ricardo Pereira da Rosa, Carlos Cauduro Schirmer, Richard Ricachenevski Gurski, João Pedro Bueno Telles, Fernando Mielke, Luíse Meurer, Maria Isabel Albano Edelweiss, Cleber Dario Pinto Kruel
ABSTRACT
To correlate the expression of p53 protein and VEGF with the prognosis of patients submitted to curative resection to treat esophageal adenocarcinoma. Forty-six patients with esophageal adenocarcinoma, submitted to curative resection, were studied. The expressions of p53 protein and VEGF were assessed by immunohistochemistry in 52.2% and 47.8% of tumors, respectively. P53 protein and VEGF expressions coincided in 26% of the cases, and no correlation between these expressions was observed. None of the clinicopathological factors showed a significant correlation with p53 protein or VEGF expressions. There was no significant association between p53 protein and VEGF expressions and long-term survival. The expression of p53 protein and VEGF did not correlate with prognosis in esophageal adenocarcinoma patients submitted to curative resection.
Sigma-Aldrich
α-Naphthyl Butyrate Solution