MilliporeSigma
  • Humoral and Cell-Mediated Immune Response in Colostrum from Women Diagnosed Positive for SARS-CoV-2.
Humoral and Cell-Mediated Immune Response in Colostrum from Women Diagnosed Positive for SARS-CoV-2.
Breastfeeding medicine : the official journal of the Academy of Breastfeeding Medicine (2021-08-13)
Vignesh Narayanaswamy, Brian Pentecost, Dominique Alfandari, Emily Chin, Kathleen Minor, Alyssa Kastrinakis, Tanya Lieberman, Kathleen F Arcaro, Heidi Leftwich
ABSTRACT
Objective: To evaluate the immune response to severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) in colostrum from women who tested positive for the virus. Methods: Between March and September 2020 we obtained bilateral colostrum samples collected on spot cards within 48 hours of delivery from 15 new mothers who had previously tested positive for SARS-CoV-2. Four of 15 women provided liquid colostrum, which was used for validating results obtained from spot cards. Archived bilateral colostrum samples collected from 8 women during 2011-2013 were used as pre-coronavirus disease 2019 (COVID-19) controls. All samples were tested for reactivity to the receptor binding domain (RBD) of the SARS-CoV-2 spike protein using an enzyme-linked immunosorbent assay that measures SARS-CoV-2 RBD-specific IgA, IgG, and IgM and for levels of 10 inflammatory cytokines (interferon-gamma [IFN-γ], tumor necrosis factor-alpha, interleukin [IL]-1β, IL-2, IL-4, IL-6, IL-8, IL-10, IL-12, IL-13) using a multiplex electrochemiluminescent sandwich assay. Results: Our validation studies indicate that the levels of SARS-CoV-2-specific antibodies and the associated cytokines measured in liquid colostrum are comparable to levels eluted from spot cards. Bilateral colostrum samples from 73%, 73%, and 33% of the 15 COVID-19 mothers exhibited IgA, IgG, and IgM reactivity to RBD, respectively. In addition, symptomatic COVID-19 mothers had statistically significant elevated levels of 4 of the 10 inflammatory markers (IFN-γ, IL-4, IL-6, and IL-12) compared to asymptomatic COVID-19 mothers. Conclusions: A strong humoral immune response is present in the colostrum of women who were infected with SARS-CoV-2 before delivering. The evolution and duration of the antibody response, as well as dynamics of the cytokine response, remain to be determined. Our results also indicate that future large-scale studies can be conducted with milk easily collected on paper spot cards.
Sigma-Aldrich
2,2′-Azino-bis(3-ethylbenzothiazoline-6-sulfonic acid) diammonium salt, tablet, 10 mg substrate per tablet