MilliporeSigma
  • Tumour gene expression signature in primary melanoma predicts long-term outcomes.
Tumour gene expression signature in primary melanoma predicts long-term outcomes.
Nature communications (2021-02-20)
Manik Garg, Dominique-Laurent Couturier, Jérémie Nsengimana, Nuno A Fonseca, Matthew Wongchenko, Yibing Yan, Martin Lauss, Göran B Jönsson, Julia Newton-Bishop, Christine Parkinson, Mark R Middleton, D Timothy Bishop, Sarah McDonald, Nikki Stefanos, John Tadross, Ismael A Vergara, Serigne Lo, Felicity Newell, James S Wilmott, John F Thompson, Georgina V Long, Richard A Scolyer, Pippa Corrie, David J Adams, Alvis Brazma, Roy Rabbie
ABSTRACT
Adjuvant systemic therapies are now routinely used following resection of stage III melanoma, however accurate prognostic information is needed to better stratify patients. We use differential expression analyses of primary tumours from 204 RNA-sequenced melanomas within a large adjuvant trial, identifying a 121 metastasis-associated gene signature. This signature strongly associated with progression-free (HR = 1.63, p = 5.24 × 10-5) and overall survival (HR = 1.61, p = 1.67 × 10-4), was validated in 175 regional lymph nodes metastasis as well as two externally ascertained datasets. The machine learning classification models trained using the signature genes performed significantly better in predicting metastases than models trained with clinical covariates (pAUROC = 7.03 × 10-4), or published prognostic signatures (pAUROC < 0.05). The signature score negatively correlated with measures of immune cell infiltration (ρ = -0.75, p < 2.2 × 10-16), with a higher score representing reduced lymphocyte infiltration and a higher 5-year risk of death in stage II melanoma. Our expression signature identifies melanoma patients at higher risk of metastases and warrants further evaluation in adjuvant clinical trials.