MilliporeSigma
  • The Influence of Bisphenol A (BPA) on the Occurrence of Selected Active Substances in Neuregulin 1 (NRG1)-Positive Enteric Neurons in the Porcine Large Intestine.
The Influence of Bisphenol A (BPA) on the Occurrence of Selected Active Substances in Neuregulin 1 (NRG1)-Positive Enteric Neurons in the Porcine Large Intestine.
International journal of molecular sciences (2021-10-14)
Krystyna Makowska, Kamila Szymańska, Jarosław Całka, Sławomir Gonkowski
ABSTRACT
Bisphenol A (BPA) is a substance used in the manufacture of plastics which shows multidirectional adverse effects on living organisms. Since the main path of intoxication with BPA is via the gastrointestinal (GI) tract, the stomach and intestine are especially vulnerable to the impact of this substance. One of the main factors participating in the regulation of intestinal functions is the enteric nervous system (ENS), which is characterized by high neurochemical diversity. Neuregulin 1 (NRG1) is one of the lesser-known active substances in the ENS. During the present study (performed using the double immunofluorescence method), the co-localization of NRG1 with other neuronal substances in the ENS of the caecum and the ascending and descending colon has been investigated under physiological conditions and after the administration of BPA. The obtained results indicate that NRG1-positive neurons also contain substance P, vasoactive intestinal polypeptide, a neuronal isoform of nitric oxide synthase and galanin and the degree of each co-localization depend on the type of enteric plexus and the particular fragment of the intestine. Moreover, it has been shown that BPA generally increases the degree of co-localization of NRG1 with other substances.
Sigma-Aldrich
Monoclonal Anti-Nitric Oxide Synthase, Universal antibody produced in mouse, clone NOS-3F7-B11-B5, ascites fluid