MilliporeSigma
  • Targeted disruption of the murine erythroid band 3 gene results in spherocytosis and severe haemolytic anaemia despite a normal membrane skeleton.
Targeted disruption of the murine erythroid band 3 gene results in spherocytosis and severe haemolytic anaemia despite a normal membrane skeleton.
Nature genetics (1996-10-01)
C D Southgate, A H Chishti, B Mitchell, S J Yi, J Palek
ABSTRACT
Band 3 is the most abundant integral protein of the red blood cell membrane. It performs two critical biological functions: maintaining ionic homeostasis, by transporting Cl- and HCO3-ions, and providing mechanical stability to the erythroid membrane. Erythroid band 3 (AE1) is one of three anion exchangers that are encoded by separate genes. The AE1 gene is transcribed by two promoters: the upstream promoter produces erythroid band 3, whereas the downstream promoter initiates transcription of the band 3 isoform in kidney. To assess the biological consequences of band 3 deficiency, we have selectively inactivated erythroid but not kidney band 3 by gene targeting in mice. Although no death in utero occurred, the majority of homozygous mice die within two weeks after birth. The erythroid band 3 null mice show retarded growth, spherocytic red blood cell morphology and severe haemolytic anaemia. Remarkably, the band 3-/- red blood cells assembled normal membrane skeleton thus challenging the notion that the presence of band 3 is required for the stable biogenesis of membrane skeleton. The availability of band 3-/- mice offers a unique opportunity to investigate the role of erythroid band 3 in the regulation of membrane-skeletal interactions, anion transport and the invasion and growth of malaria parasite into red blood cells.