MilliporeSigma
  • Effects of acute inflammation on plasma retinol, retinol-binding protein, and its mRNA in the liver and kidneys of vitamin A-sufficient rats.
Effects of acute inflammation on plasma retinol, retinol-binding protein, and its mRNA in the liver and kidneys of vitamin A-sufficient rats.
Journal of lipid research (1996-05-01)
F J Rosales, S J Ritter, R Zolfaghari, J E Smith, A C Ross
ABSTRACT
The acute inflammatory response to tissue injury and infection is associated with low concentrations of plasma retinol and its specific transport proteins, retinol-binding protein (RBP) and transthyretin (TTR). To examine the kinetics and mechanism of hyporetinemia, we have induced acute inflammation with lipopolysaccharide (LPS, from Pseudomonas aeruginosa) in rats with adequate stores of vitamin A. Twenty-four h after treatment with LPS (50 micrograms i.p. per 100 g body weight) or saline and food withdrawal, plasma retinol equalled 0.72 +/- 0.06 mumol/L (mean +/- SEM) in five LPS-treated rats versus 1.35 +/- 0.1 mumol/L in five saline-treated rats (P < 0.01). Plasma, liver, and kidney RBP and TTR concentrations were also significantly reduced, but liver and kidney retinol concentrations did not differ between treatment groups. The relative abundance of RBP mRNA in liver (LPS treatment compared to saline treatment) was reduced as early as 12 h (0.44 +/- 0.15, n = 4 pairs, P < 0.02), and continued to be reduced at 24 h (0.57 +/- 0.12, n = 5 pairs, P < 0.02). In the kidney this ratio did not change significantly due to LPS treatment. The relative abundance of cellular retinol-binding protein (CRBP) mRNA in liver and kidney also was not affected by LPS treatment. We infer from these data that inflammation-induced hyporetinemia results from a reduction in the hepatic synthesis of RBP and secretion of the retinol-RBP complex. Moreover, the results imply that plasma retinol concentration is a poor indicator of vitamin A status during inflammation.
Sigma-Aldrich
Retinol Binding Protein from human urine, buffered aqueous solution
Pricing and availability is not currently available.