MilliporeSigma
  • Bcl-2 sensitivity differentiates two pathways for motoneuronal death in the wobbler mutant mouse.
Bcl-2 sensitivity differentiates two pathways for motoneuronal death in the wobbler mutant mouse.
The Journal of neuroscience : the official journal of the Society for Neuroscience (1996-10-01)
M Coulpier, M P Junier, M Peschanski, P A Dreyfus
ABSTRACT
The molecular events leading to motoneuronal death are still poorly understood. In mammals, the bcl-2 proto-oncogene, which encodes a membrane-associated protein, has been shown to suppress both developmental motoneuronal death and experimental axotomy-induced motoneuronal death. We assessed a potential protective effect of Bcl-2 on pathological motoneuronal death processes in adult rodents. We took advantage of the murine mutant wobbler, which undergoes progressive degeneration of the spinal and brainstem motoneurons. A hybrid carrying both the wobbler mutation and the human bcl-2 transgene under the control of the neuron-specific enolase promoter was produced. Although Bcl-2 protected spinal and brainstem motoneurons from developmental death and the postnatal motoneurons of the facial nucleus from axotomy-induced death, the pathological motoneuronal death was not altered in the adult hybrid. These results demonstrate that Bcl-2 sensitivity distinguishes at least two different motoneuronal death pathways in the wobbler mutant. They support the hypothesis that experimental and pathological motoneuronal death are dependent on different cellular mechanisms.