MilliporeSigma
  • Inhibition of growth of OV-1063 human epithelial ovarian cancer xenografts in nude mice by treatment with luteinizing hormone-releasing hormone antagonist SB-75.
Inhibition of growth of OV-1063 human epithelial ovarian cancer xenografts in nude mice by treatment with luteinizing hormone-releasing hormone antagonist SB-75.
Proceedings of the National Academy of Sciences of the United States of America (1994-07-19)
T Yano, J Pinski, G Halmos, K Szepeshazi, K Groot, A V Schally
ABSTRACT
Female athymic nude mice bearing xenografts of OV-1063 human epithelial ovarian cancer cell line were treated with potent luteinizing hormone (LH)-releasing hormone (LH-RH) antagonist SB-75 (Cetrorelix; [Ac-D-Nal(2)1, D-Phe(4 CI)2, D-Pal(3)3, D-Cit6, D-Ala10]LH-RH in which Ac-D-Nal(2) = N-acetyl-3-(2-naphthyl)-D-alanine, D-Phe(4CI) = 4-chloro-D-phenylalanine, D-Pal(3) = 3-(3-pyridyl)-D-alanine, and D-Cit = D-Citrulline) or with the agonist [D-Trp6]LH-RH. In the first experiment, SB-75 and [D-Trp6]LH-RH were administered in the form of microcapsules releasing 60 and 25 micrograms/day, respectively. In the second study, the analogs were given by daily s.c. injections in doses of 100 micrograms/day. In both experiments, tumor growth, as measured by reduction in tumor volume, percentage change in tumor volume, tumor burden, and increase in tumor doubling time, was significantly inhibited by treatment with SB-75 but not with [D-Trp6]LH-RH. Uterine and ovarian weights were reduced and serum LH levels decreased by administration of either analog. Chronic treatment with SB-75 greatly reduced the concentration of receptors for epidermal growth factor and insulin-like growth factor I in tumor cell membranes, a phenomenon that might be related to tumor growth inhibition. It is possible that the antitumoral effects of SB-75 on OV-1063 ovarian cancers are exerted not only through the suppression of the pituitary-gonadal axis, but also directly. In view of its strong inhibitory effect on the growth of OV-1063 ovarian cancers in vivo, the potent LH-RH antagonist SB-75 might be considered for possible hormonal therapy of advanced epithelial ovarian carcinoma.