MilliporeSigma
  • The isolation and characterization of differentially phosphorylated fractions of phosphofructokinase from rabbit skeletal muscle.
The isolation and characterization of differentially phosphorylated fractions of phosphofructokinase from rabbit skeletal muscle.
European journal of biochemistry (1977-11-01)
C R Hussey, P F Liddle, D Ardron, G L Kellett
PMID144600
ABSTRACT
A preparation of phosphofructokinase from rabbit skeletal muscle is described which exploits the association-dissociation properties of the enzyme. Phosphofructokinase to prepared is partially phosphorylated and may be fractioned into three distinct species with sedimentation coefficients of 30 S, 18 S and 13 S by chromatography of agarose gels, hydroxyapatite or DEAE-cellulose. Measurements of alkali-labile phosphate content (phosphoserine and/or phosphothreonine) show that fractions consisting almost exclusively of 30-S species and fractions consisting predominantly of 18-S and 13-S species contain approximately 0.15 and 0.29 mol of phosphate per phosphofructokinase monomer (Mr = 80000) respectively. The results are interpreted in terms of at least two 13-S components which differ in their phosphate contents and also in their self-association properties. The possible significance of phosphorylation is discussed.
Sigma-Aldrich
α-Glycerophosphate Dehydrogenase-Triosephosphate Isomerase from rabbit muscle, Type III, ammonium sulfate suspension