MilliporeSigma
  • Squamous cell carcinomas. An immunohistochemical study of cytokeratins and involucrin in primary and metastatic tumours.
Squamous cell carcinomas. An immunohistochemical study of cytokeratins and involucrin in primary and metastatic tumours.
Histopathology (1993-07-01)
Z Suo, R Holm, J M Nesland
ABSTRACT
The expression of cytokeratins (CK) 1, 4, 5/6, 8, 13, 18, 19 and 20 and involucrin in 42 cases of squamous cell carcinomas from various locations was examined. The tumours expressed CK5/6 in 55%, CK8 in 76%, CK13 in 43% and CK19 in 95% of cases. The CK5/6-positive primary tumours were from uterine cervix, head and neck, lung, skin, oesophagus and urinary bladder, and the CK13-positive primary tumours were from uterine cervix, lung and vulva. Metastatic squamous cell carcinomas from head and neck more frequently expressed CK5/6 and 13, 7/7 (100%) and 6/7 (86%) compared with 3/5 (60%) and 0/5 (0%) in the primary squamous cell carcinomas. Few cases were CK1, CK4 and CK18 immunoreactive. CK20 immunoreactivity was not observed. Involucrin was expressed in 71% of tumours, and most of the involucrin-positive cells were located at the central parts of tumour cell clusters except for one case in which the peripheral cells around tumour cell clusters were positive. Thus, expression of the so-called simple epithelial markers CK8 and CK19 occurs in the majority of squamous cell carcinomas. The absence of CK20 immunoreactivity may be helpful in differential diagnosis.