MilliporeSigma
  • Effect of a Phytogenic Feed Additive on Growth Performance, Nutrient Digestion, and Immune Response in Broiler-Fed Diets with Two Different Levels of Crude Protein.
Effect of a Phytogenic Feed Additive on Growth Performance, Nutrient Digestion, and Immune Response in Broiler-Fed Diets with Two Different Levels of Crude Protein.
Animals : an open access journal from MDPI (2021-04-04)
Jinquan Wang, Shengchen Su, Chasity Pender, Raj Murugesan, Basharat Syed, Woo Kyun Kim
ABSTRACT
The aim of this experiment was to evaluate the effect of a phytogenic feed additive (PFA) on growth performance and nutrient digestibility of broilers fed corn and soybean meal-based diets containing two different levels of crude protein. A 2 × 2 completely randomized factorial arrangement (eight replicates/treatment, 30 birds/replicate) was conducted with a positive control (PC) and negative control (NC) containing crude protein at standard or reduced by 1.5% (equivalent to a reduction of 15 g/kg), respectively, and supplementation of PFA at 0 or 125 ppm of diet. There were no significant interactions found between PFA and CP levels in the current study. Main effect analysis showed that during 0-42 d of age NC diets decreased body weight gain (p < 0.05), but increased feed intake (p < 0.05) and feed conversion ratio (FCR, p < 0.01), whereas supplementation of PFA resulted in a lower FCR (p < 0.01). The ileal nutrient digestibility was reduced (p < 0.05) in the broilers fed a reduced protein diet at 21 d compared to the standard protein level group, but there were no effects for PFA levels. Similarly, supplementing PFAs showed no effects on digestive enzyme (Alkaline phosphatase, amylase, and lipase) activity in jejunal digesta and jejunal brush border enzyme (maltase, sucrase, and aminopeptidase) activity. Supplementation of PFA downregulated (p < 0.05) the mRNA expressions of cytochrome P450 1A and interleukin 6 in the ileum but had no effects on nutrient transporter genes in the jejunum. In conclusion, supplementation of PFA reduced broiler FCR during the whole grow-out period and positively regulated the immune responses in the ileum.