MilliporeSigma
  • Optimization of a process for the production of (R)-2-hydroxy-4-phenylbutyric acid--an intermediate for inhibitors of angiotensin converting enzyme.
Optimization of a process for the production of (R)-2-hydroxy-4-phenylbutyric acid--an intermediate for inhibitors of angiotensin converting enzyme.
Journal of biotechnology (1992-07-01)
E Schmidt, O Ghisalba, D Gygax, G Sedelmeier
ABSTRACT
(R)-2-Hydroxy-4-phenylbutyric acid, an intermediate in the manufacture of inhibitors of angiotensin converting enzyme, can be produced continuously in an enzyme membrane reactor by enzymatic reduction of its corresponding alpha-keto acid. D-Lactate dehydrogenase (D-LDH) from Staphylococcus epidermidis was chosen as the most appropriate enzyme to carry out the NADH-dependent reduction. Formate dehydrogenase (FDH) was used for NADH regeneration. Detailed kinetic measurements and a mathematical model for the coupled enzyme reactions were applied to calculate the optimal conditions for continuous production of the alpha-hydroxy acid. A mass of 1 kg [corrected] (R)-2-hydroxy-4-phenylbutyric acid was synthesized in a 220 ml enzyme membrane reactor over a period of 4 weeks. A mean space-time-yield of 165 g l-1 d-1 was achieved at low enzyme consumptions of 150 U kg-1 alpha-hydroxy acid for FDH and D-LDH.
Sigma-Aldrich
D-Lactic Dehydrogenase from Staphylococcus epidermidis, lyophilized powder, ≥80 units/mg solid