MilliporeSigma
  • Identification of the 64K autoantigen in insulin-dependent diabetes as the GABA-synthesizing enzyme glutamic acid decarboxylase.
Identification of the 64K autoantigen in insulin-dependent diabetes as the GABA-synthesizing enzyme glutamic acid decarboxylase.
Nature (1990-09-13)
S Baekkeskov, H J Aanstoot, S Christgau, A Reetz, M Solimena, M Cascalho, F Folli, H Richter-Olesen, P De Camilli, P D Camilli
ABSTRACT
The pancreatic islet beta-cell autoantigen of relative molecular mass 64,000 (64K), which is a major target of autoantibodies associated with the development of insulin-dependent diabetes mellitus (IDDM) has been identified as glutamic acid decarboxylase, the biosynthesizing enzyme of the inhibitory neurotransmitter GABA (gamma-aminobutyric acid). Pancreatic beta cells and a subpopulation of central nervous system neurons express high levels of this enzyme. Autoantibodies against glutamic acid decarboxylase with a higher titre and increased epitope recognition compared with those usually associated with IDDM are found in stiff-man syndrome, a rare neurological disorder characterized by a high coincidence with IDDM.