MilliporeSigma
  • Retinoic acid rewires the adrenergic core regulatory circuitry of childhood neuroblastoma.
Retinoic acid rewires the adrenergic core regulatory circuitry of childhood neuroblastoma.
Science advances (2021-10-21)
Mark W Zimmerman, Adam D Durbin, Shuning He, Felix Oppel, Hui Shi, Ting Tao, Zhaodong Li, Alla Berezovskaya, Yu Liu, Jinghui Zhang, Richard A Young, Brian J Abraham, A Thomas Look
ABSTRACT
[Figure: see text].
Sigma-Aldrich
Anti-PCNA Antibody, clone PC10, clone PC10, Chemicon®, from mouse
Pricing and availability is not currently available.