MilliporeSigma
Human placental alkaline phosphatase in liver and intestine.
Proceedings of the National Academy of Sciences of the United States of America (1985-09-01)
E Garattini, J Margolis, E Heimer, A Felix, S Udenfriend
ABSTRACT
Three distinct forms of human alkaline phosphatase, presumably isozymes, are known, each apparently associated with a specific tissue. These are placental, intestinal, and liver (kidney and bone). We have used a specific immunoassay and HPLC to show that placental alkaline phosphatase is also present in extracts of liver and intestine in appreciable amounts.