MilliporeSigma
  • A cross-talk between CAR T cell subsets and the tumor microenvironment is essential for sustained cytotoxic activity.
A cross-talk between CAR T cell subsets and the tumor microenvironment is essential for sustained cytotoxic activity.
Science immunology (2021-03-28)
Morgane Boulch, Marine Cazaux, Yann Loe-Mie, Ronan Thibaut, Béatrice Corre, Fabrice Lemaître, Capucine L Grandjean, Zacarias Garcia, Philippe Bousso
ABSTRACT
Chimeric antigen receptor (CAR) T cell therapy relies on the activity of a large pool of tumor-targeting cytotoxic effectors. Whether CAR T cells act autonomously or require interactions with the tumor microenvironment (TME) remains incompletely understood. Here, we report an essential cross-talk between CAR T cell subsets and the TME for tumor control in an immunocompetent mouse B cell lymphoma model of anti-CD19 CAR T cell therapy. Using single-cell RNA sequencing, we revealed substantial modification of the TME during CAR T cell therapy. Interferon-γ (IFN-γ) produced by CAR T cells not only enhanced endogenous T and natural killer cell activity but was also essential for sustaining CAR T cell cytotoxicity, as revealed by intravital imaging. CAR T cell-derived IFN-γ facilitated host interleukin-12 production that supported host immune and CAR T cell responses. Compared with CD8+ CAR T cells, CD4+ CAR T cells were more efficient at host immune activation but less capable of direct tumor killing. In summary, CAR T cells do not act independently in vivo but rely instead on cytokine-mediated cross-talk with the TME for optimal activity. Invigorating CAR T cell interplay with the host represents an attractive strategy to prevent relapses after therapy.
Sigma-Aldrich
Interleukin-12 from mouse, ≥97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.