MilliporeSigma
  • Peptide sweeteners. 6. Structural studies on the C-terminal amino acid of L-aspartyl dipeptide sweeteners.
Peptide sweeteners. 6. Structural studies on the C-terminal amino acid of L-aspartyl dipeptide sweeteners.
Journal of medicinal chemistry (1984-12-01)
J W Tsang, B Schmied, R Nyfeler, M Goodman
ABSTRACT
Stereochemical and structural aspects of the variations in the C-terminal residue of L-aspartyl-L-phenylalanine methyl ester have been investigated. Novel configurational analogues such as L-aspartyl-D-alanine benzyl ester and L-aspartyl-D-alpha-aminobutyric acid benzyl ester were found to be sweet. In addition, chiral and achiral alpha, alpha-dialkylglycine and alpha-aminocycloalkanecarboxylic acids were incorporated into the dipeptides. The L-aspartic acid based dipeptide derivatives of alpha-aminoisobutyric acid methyl ester, alpha-aminocyclopropanecarboxylic acid methyl ester, alpha-aminocyclobutanecarboxylic acid methyl ester, and alpha-aminocyclopentanecarboxylic acid methyl ester are sweet. Dipeptides with alpha-aminocyclohexanecarboxylic acid methyl ester and alpha-aminocycloheptanecarboxylic acid methyl ester are bitter, whereas the analogues with alpha-aminocyclooctanecarboxylic acid methyl ester, alpha, alpha-diethylglycine methyl ester, and alpha-aminoisobutyric acid benzyl ester are tasteless. Aspects on chirality and effective volume of the C-terminal residue are discussed and correlated with taste.