MilliporeSigma
  • Stereospecificity of sodium borohydride reduction of tyrosine decarboxylase from Streptococcus faecalis.
Stereospecificity of sodium borohydride reduction of tyrosine decarboxylase from Streptococcus faecalis.
The Journal of biological chemistry (1979-06-25)
J C Vederas, I D Reingold, H W Sellers
PMID36380
ABSTRACT
Sodium boro[3H]hydride reduction of tyrosine decarboxylase from Streptococcus faecalis followed by complete hydrolysis of the enzyme produces epsilon-[3H]pyridoxyllysine. Degradation of this material to [4'-3H]pyridoxamine and stereochemical analysis with apoaspartate aminotransferase shows that the re side at C-4' of the cofactor is exposed to solvent at pH 5.5 and 7.0. After binding of L-tyrosine at pH 5.5 or tyramine at pH 7.0 to the holoenzyme, sodium boro[3H]hydride reduction proceeds from the si face at C-4' of the substrate . cofactor complex. This indicates one of two conformational changes occurs upon binding of substrate; either rotation about the C-4 to C-4' bond in the cofactor or rotation about the axis through the C-5 and C-5' bond.
Sigma-Aldrich
L-Tyrosine Decarboxylase Apoenzyme from Streptococcus faecalis, <0.005 unit/mg solid (without pyridoxal 5-phosphate), ≥0.05 unit/mg solid (with excess pyridoxal 5-phosphate)