MilliporeSigma
  • Production of L-arginine by arginine hydroxamate-resistant mutants of Bacillus subtilis.
Production of L-arginine by arginine hydroxamate-resistant mutants of Bacillus subtilis.
Applied microbiology (1971-12-01)
M Kisumi, J Kato, M Sugiura, I Chibata
ABSTRACT
l-Arginine hydroxamate inhibited the growth of various bacteria, and the inhibition was readily reversed by arginine. l-Arginine hydroxamate (10(-3)m) completely inhibited the growth of Bacillus subtilis. This inhibitory effect was prevented by 2.5 x 10(-4)ml-arginine, which was the most effective of all the natural amino acids in reversing the inhibition. l-Arginine hydroxamate-resistant mutants of Bacillus subtilis were isolated and found to excrete l-arginine in relatively high yields. One of the mutants, strain AHr-5, produced 4.5 mg of l-arginine per ml in shaken culture in 3 days.