MilliporeSigma
  • Systematic Proteome and Lysine Succinylome Analysis Reveals Enhanced Cell Migration by Hyposuccinylation in Esophageal Squamous Cell Carcinoma.
Systematic Proteome and Lysine Succinylome Analysis Reveals Enhanced Cell Migration by Hyposuccinylation in Esophageal Squamous Cell Carcinoma.
Molecular & cellular proteomics : MCP (2021-02-10)
Zhenchang Guo, Feng Pan, Liu Peng, Shanshan Tian, Jiwei Jiao, Liandi Liao, Congcong Lu, Guijin Zhai, Zhiyong Wu, Hanyang Dong, Xiue Xu, Jianyi Wu, Pu Chen, Xue Bai, Dechen Lin, Liyan Xu, Enmin Li, Kai Zhang
ABSTRACT
Esophageal squamous cell carcinoma (ESCC) is an aggressive malignancy with poor therapeutic outcomes. However, the alterations in proteins and posttranslational modifications (PTMs) leading to the pathogenesis of ESCC remain unclear. Here, we provide the comprehensive characterization of the proteome, phosphorylome, lysine acetylome, and succinylome for ESCC and matched control cells using quantitative proteomic approach. We identify abnormal protein and PTM pathways, including significantly downregulated lysine succinylation sites in cancer cells. Focusing on hyposuccinylation, we reveal that this altered PTM was enriched on enzymes of metabolic pathways inextricably linked with cancer metabolism. Importantly, ESCC malignant behaviors such as cell migration are inhibited once the level of succinylation was restored in vitro or in vivo. This effect was further verified by mutations to disrupt succinylation sites in candidate proteins. Meanwhile, we found that succinylation has a negative regulatory effect on histone methylation to promote cancer migration. Finally, hyposuccinylation is confirmed in primary ESCC specimens. Our findings together demonstrate that lysine succinylation may alter ESCC metabolism and migration, providing new insights into the functional significance of PTM in cancer biology.
Sigma-Aldrich
Aldolase Activity Colorimetric Assay Kit, Sufficient for 100 Colorimetric tests