MilliporeSigma
  • [Phospholipase A2 receptor antigen antibodies in serum and renal biopsies of patients with membranous nephropathy].
[Phospholipase A2 receptor antigen antibodies in serum and renal biopsies of patients with membranous nephropathy].
Revista medica de Chile (2021-06-10)
Alondra Frías Oyarzún, Paula Segura Hidalgo, Luis Toro Cabrera, Leticia Elgueta Segura, Rubén Torres Díaz, Miriam Alvo Abodovsky
ABSTRACT
The discovery of the phospholipase A2 receptor antigen and its highly specific autoantibody (anti-PLA2R Ab) was useful for the diagnosis and follow-up of patients with membranous nephropathy (MN). Thus, some international guidelines recommend not performing renal biopsy in patients with positive serum anti-PLA2R Ab. To evaluate the prevalence of anti-PLA2R Ab in serum and renal tissue samples from Chilean patients with primary MN. Twenty-eight patients aged 50 ± 14 years (20 males) with biopsy-proven primary MN plus a negative workup for secondary causes were included. Measurements of serum and renal histologic anti-PLA2R Ab were performed. The relationship between the findings of serum and tissue anti-PLA2R Ab was evaluated. Fifteen patients (54 %) had anti-PLA2R Ab presence in serum and 19 patients (68%) had positive anti-PLA2R Ab in the renal biopsy. All patients with positive serum anti-PLA2R Ab had positive antibodies on immunohistochemistry. Serum anti-PLA2R Ab is potentially useful in the diagnosis of patients with suspected primary MN in Chilean population.
Sigma-Aldrich
Anti-PLA2R1 antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution