MilliporeSigma
  • Loss of complex O-glycosylation impairs exocrine pancreatic function and induces MODY8-like diabetes in mice.
Loss of complex O-glycosylation impairs exocrine pancreatic function and induces MODY8-like diabetes in mice.
Experimental & molecular medicine (2018-10-12)
Gerrit Wolters-Eisfeld, Baris Mercanoglu, Bianca T Hofmann, Thomas Wolpers, Claudia Schnabel, Sönke Harder, Pascal Steffen, Kai Bachmann, Babett Steglich, Jörg Schrader, Nicola Gagliani, Hartmut Schlüter, Cenap Güngör, Jakob R Izbicki, Christoph Wagener, Maximilian Bockhorn
ABSTRACT
Cosmc is ubiquitously expressed and acts as a specific molecular chaperone assisting the folding and stability of core 1 synthase. Thus, it plays a crucial role in the biosynthesis of O-linked glycosylation of proteins. Here, we show that ablation of Cosmc in the exocrine pancreas of mice causes expression of truncated O-glycans (Tn antigen), resulting in exocrine pancreatic insufficiency with decreased activities of digestive enzymes and diabetes. To understand the molecular causes of the pleiotropic phenotype, we used Vicia villosa agglutinin to enrich Tn antigen-modified proteins from Cosmc-KO pancreatic lysates and performed a proteomic analysis. Interestingly, a variety of proteins were identified, of which bile salt-activated lipase (also denoted carboxyl-ester lipase, Cel) was the most abundant. In humans, frameshift mutations in CEL cause maturity-onset diabetes of the young type 8 (MODY8), a monogenic syndrome of diabetes and pancreatic exocrine dysfunction. Here, we provide data suggesting that differentially O-glycosylated Cel could negatively affect beta cell function. Taken together, our findings demonstrate the importance of correct O-glycan formation for normal exocrine and endocrine pancreatic function, implying that aberrant O-glycans might be relevant for pathogenic mechanisms of the pancreas.
Sigma-Aldrich
N-Methoxysuccinyl-Ala-Ala-Pro-Val-7-amido-4-methylcoumarin, elastase substrate
Pricing and availability is not currently available.