MilliporeSigma
  • Oestrogenic activity of CPRG (chlorophenol red-β-D-galactopyranoside), a β-galactosidase substrate commonly used in recombinant yeast oestrogenic assays.
Oestrogenic activity of CPRG (chlorophenol red-β-D-galactopyranoside), a β-galactosidase substrate commonly used in recombinant yeast oestrogenic assays.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals (2001-01-01)
Els Vanderperren Wim Demaré Ronny Blust Kris Cooreman Peter Bossier
ABSTRACT
The finding that a variety of chemicals display oestrogenic activity has resulted in the development of in vitro and in vivo assays to assess oestrogenic activity. One such assay, the yeast oestrogen assay (YES) makes use of recombinant yeast cells that harbour an oestrogen receptor expression cassette and a reporter construct, coding for bgalactosidase. The induction mechanism starts with the binding of oestrogenic compounds to the oestrogen receptor. This complex activates the production of β-galactosidase. The β-galactosidase activity is thus a measure of the oestrogenic activity of chemical compounds. In the YES assay, the β-galactosidase activity may be quantified with the chromogenic substrate chlorophenol red-β-d-galactopyranoside (CPRG). In the present study it is reported that CPRG or its β-galactosidase degradation product chlorophenol red act in the YES as an oestrogenic compound itself. The implications of this finding are described. It is especially argued that chlorophenol red production after prolonged incubation of the assay might be misinterpreted as an oestrogenic effect of the test compound.
Product Description
Roche
CPRG, Chlorophenol red-β-D-galactopyranoside