MilliporeSigma
  • Michael addition of pyrimidine with disaccharide acrylates catalyzed in organic medium with lipase M from Mucor javanicus.
Michael addition of pyrimidine with disaccharide acrylates catalyzed in organic medium with lipase M from Mucor javanicus.
Biotechnology letters (2006-09-07)
Hong-Xia Dai, Shi-Ping Yao, Jian Wang
ABSTRACT
The Michael addition of fluorouracil (0.01 mol), uracil (0.01 mol), thymine (0.01 mol) to disaccharide acrylates (0.03 mol) was carried out in pyridine (20 ml) at 50 degrees C for 72 h with lipase M from Mucor javanicus (200 mg). Six new pyrimidine derivatives containing a branched sugar were obtained in yields from 56 to 75%.
Sigma-Aldrich
Amano Lipase M from Mucor javanicus, ≥10,000 U/g, pH 7.0, 40 °C (Optimum pH and temperature)