MilliporeSigma
  • ZM241385, DPCPX, MRS1706 are inverse agonists with different relative intrinsic efficacies on constitutively active mutants of the human adenosine A2B receptor.
ZM241385, DPCPX, MRS1706 are inverse agonists with different relative intrinsic efficacies on constitutively active mutants of the human adenosine A2B receptor.
The Journal of pharmacology and experimental therapeutics (2006-11-02)
Qilan Li, Kai Ye, Clara C Blad, Hans den Dulk, Jaap Brouwer, Ad P Ijzerman, Margot W Beukers
ABSTRACT
The human adenosine A(2B) receptor belongs to class A G protein-coupled receptors (GPCRs). In our previous work, constitutively active mutant (CAM) human adenosine A(2B) receptors were identified from a random mutation bank. In the current study, three known A(2B) receptor antagonists, 4-{2-[7-amino-2-(2-furyl)[1,2,4]triazolo-[2,3-a][1,3,5]triazin-5-yl-amino]ethyl}phenol (ZM241385), 8-cyclopentyl-1,3-dipropylxanthine (DPCPX), and N-(4-acetylphenyl)-2-[4-(2,3,6,7-tetrahydro-2,6-dioxo-1,3-dipropyl-1H-purin-8-yl)phenoxy]acetamide (MRS1706) were tested on wild-type and nine CAM A(2B) receptors with different levels of constitutive activity in a yeast growth assay. All three compounds turned out to be inverse agonists for the adenosine A(2B) receptor because they were able to fully reverse the basal activity of four low-level constitutively active A(2B) receptor mutants and to partially reverse the basal activity of three medium-level constitutively active A(2B) receptor mutants. We also discovered two highly constitutively active mutants whose basal activity could not be reversed by any of the three compounds. A two-state receptor model was used to explain the experimental observations; fitting these yielded the following relative intrinsic efficacies for the three inverse agonists ZM241385, DPCPX, and MRS1706: 0.14 +/- 0.03, 0.35 +/- 0.03, and 0.31 +/- 0.02, respectively. Moreover, varying L, the ratio of active versus inactive receptors in this model, from 0.11 for mutant F84L to 999 for two highly constitutively active mutants yielded simulated dose-response curves that mimicked the experimental curves. This study is the first description of inverse agonists for the human adenosine A(2B) receptor. Moreover, the use of receptor mutants with varying levels of constitutive activity enabled us to determine the relative intrinsic efficacy of these inverse agonists.
Sigma-Aldrich
MRS-1706, ≥98% (HPLC)
Pricing and availability is not currently available.