MilliporeSigma
  • Identification of the V factor needed for synthesis of the iron-molybdenum cofactor of nitrogenase as homocitrate.
Identification of the V factor needed for synthesis of the iron-molybdenum cofactor of nitrogenase as homocitrate.
Nature (1987-10-04)
T R Hoover, A D Robertson, R L Cerny, R N Hayes, J Imperial, V K Shah, P W Ludden
ABSTRACT
Nitrogenase catalyses the ATP-dependent reduction of N2 to NH3, and is composed of two proteins, dinitrogenase (MoFe protein or component I) and dinitrogenase reductase (Fe protein or component II). Dinitrogenase contains a unique prosthetic group (iron-molybdenum cofactor, FeMoco) comprised of Fe, Mo and S, which has been proposed as the site of N2 reduction. Biochemical and genetic studies of Nif- (nitrogen fixation) mutants of Klebsiella pneumoniae which are defective in nitrogen fixation, have shown that the nifB, nifQ, nifN, nifE and nifV genes are required for the biosynthesis of FeMo-co. Recently, a system for in vitro synthesis of FeMoco was described. The assay requires at least the nifB, nifN and nifE gene products, and a low-molecular-weight factor (V factor) produced in the presence of the nifV gene product. We have used this system to study FeMoco biosynthesis. We report here the isolation of V factor and identify it as homocitric acid ([R]2-hydroxy-1,2,4-butanetricarboxylic acid).
Sigma-Aldrich
Sodium (±)-homocitrate tribasic