MilliporeSigma
  • NUDT15-mediated hydrolysis limits the efficacy of anti-HCMV drug ganciclovir.
NUDT15-mediated hydrolysis limits the efficacy of anti-HCMV drug ganciclovir.
Cell chemical biology (2021-07-01)
Si Min Zhang, Daniel Rehling, Ann-Sofie Jemth, Adam Throup, Natalia Landázuri, Ingrid Almlöf, Mona Göttmann, Nicholas C K Valerie, Sanjay R Borhade, Prasad Wakchaure, Brent D G Page, Matthieu Desroses, Evert J Homan, Martin Scobie, Sean G Rudd, Ulrika Warpman Berglund, Cecilia Söderberg-Nauclér, Pål Stenmark, Thomas Helleday
ABSTRACT
Ganciclovir (GCV) is the first-line therapy against human cytomegalovirus (HCMV), a widespread infection that is particularly dangerous for immunodeficient individuals. Closely resembling deoxyguanosine triphosphate, the tri-phosphorylated metabolite of GCV (GCV-TP) is preferentially incorporated by the viral DNA polymerase, thereby terminating chain extension and, eventually, viral replication. However, the treatment outcome of GCV varies greatly among individuals, therefore warranting better understanding of its metabolism. Here we show that NUDT15, a Nudix hydrolase known to metabolize thiopurine triphosphates, can similarly hydrolyze GCV-TP through biochemical studies and co-crystallization of the NUDT15/GCV-TP complex. More critically, GCV efficacy was potentiated in HCMV-infected cells following NUDT15 depletion by RNAi or inhibition by an in-house-developed, nanomolar NUDT15 inhibitor, TH8321, suggesting that pharmacological targeting of NUDT15 is a possible avenue to improve existing anti-HCMV regimens. Collectively, the data further implicate NUDT15 as a broad-spectrum metabolic regulator of nucleoside analog therapeutics, such as thiopurines and GCV.
Sigma-Aldrich
Pyrophosphatase, Inorganic from Escherichia coli, recombinant, expressed in E. coli, lyophilized powder, ≥90%, ≥800 units/mg protein
Sigma-Aldrich
2′-Deoxyguanosine 5′-triphosphate sodium salt hydrate, ≥96% (HPLC)