MilliporeSigma
  • The Effect of an Adsorbent Matrix on Recovery of Microorganisms from Hydrocarbon-Contaminated Groundwater.
The Effect of an Adsorbent Matrix on Recovery of Microorganisms from Hydrocarbon-Contaminated Groundwater.
Microorganisms (2021-01-07)
Nicole M Taylor, Courtney R A Toth, Victoria Collins, Paolo Mussone, Lisa M Gieg
ABSTRACT
The microbial degradation of recalcitrant hydrocarbons is an important process that can contribute to the remediation of oil and gas-contaminated environments. Due to the complex structure of subsurface terrestrial environments, it is important to identify the microbial communities that may be contributing to biodegradation processes, along with their abilities to metabolize different hydrocarbons in situ. In this study, a variety of adsorbent materials were assessed for their ability to trap both hydrocarbons and microorganisms in contaminated groundwater. Of the materials tested, a porous polymer resin (Tenax-TA) recovered the highest diversity of microbial taxa in preliminary experiments and was selected for additional (microcosm-based) testing. Oxic and anoxic experiments were prepared with groundwater collected from a contaminated aquifer to assess the ability of Tenax-TA to adsorb two environmental hydrocarbon contaminants of interest (toluene and benzene) while simultaneously providing a surface for microbial growth and hydrocarbon biodegradation. Microorganisms in oxic microcosms completely degraded both targets within 14 days of incubation, while anoxically-incubated microorganisms metabolized toluene but not benzene in less than 80 days. Community analysis of Tenax-TA-associated microorganisms revealed taxa highly enriched in sessile hydrocarbon-degrading treatments, including Saprospiraceae, Azoarcus, and Desulfoprunum, which may facilitate hydrocarbon degradation. This study showed that Tenax-TA can be used as a matrix to effectively trap both microorganisms and hydrocarbons in contaminated environmental systems for assessing and studying hydrocarbon-degrading microorganisms of interest.
Sigma-Aldrich
Benzene, anhydrous, 99.8%