MilliporeSigma
  • Spy chemistry-enabled protein directional immobilization and protein purification.
Spy chemistry-enabled protein directional immobilization and protein purification.
Biotechnology and bioengineering (2020-06-17)
Zhanglin Lin, Qiao Lin, Jiahui Li, Marco Pistolozzi, Lei Zhao, Xiaofeng Yang, Yanrui Ye
ABSTRACT
Site-directed protein immobilization allows the homogeneous orientation of proteins with high retention of activity, which is advantageous for many applications. Here, we report a facile, specific, and efficient strategy based on the SpyTag-SpyCatcher chemistry. Two SpyTag-fused model proteins, that is, the monomeric red fluorescent protein (RFP) and the oligomeric glutaryl-7-aminocephalosporanic acid acylase, were easily immobilized onto a SpyCatcher-modified resin directly from cell lysates, with activity recoveries in the range of 85-91%. This strategy was further adapted to protein purification, which proceeded through the selective capture of the SpyCatcher-fused target proteins by a SpyTag-modified resin, with the aid of an intein to generate authentic N-termini. For two model proteins, that is, RFP and a variable domain of a heavy chain antibody, the yields were ∼3-7 mg/L culture with >90% purities. This approach could provide a versatile tool for producing high-performance immobilized protein devices and proteins for industrial and therapeutic uses.
Sigma-Aldrich
7-Aminocephalosporanic acid, 98%