MilliporeSigma
  • Serum angiopoietin-like 3 levels are elevated in obese non diabetic men but are unaffected during an oral glucose tolerance test.
Serum angiopoietin-like 3 levels are elevated in obese non diabetic men but are unaffected during an oral glucose tolerance test.
Scientific reports (2020-12-05)
Maria Fernanda Garcés, Julieth Daniela Buell-Acosta, Haiver Antonio Rodríguez-Navarro, Estefania Pulido-Sánchez, Juan José Rincon-Ramírez, Diana Carolina Moreno-Ordóñez, Roberto Franco-Vega, Jhoan Sebastian Roncancio-Muñoz, Alvaro Javier Burgos-Cardenas, Ezequiel Lacunza, Justo P Castaño, Carlos Diéguez, Rubén Nogueiras, Elizabeth Sanchez, Jorge Eduardo Caminos
ABSTRACT
This study aimed to determine ANGPTL3 serum levels in healthy young lean and obese non-diabetic men during an oral glucose tolerance test (OGTT) and correlate them with anthropometric, biochemical and hormonal parameters. A case-control study was carried out and 30 young obese non-diabetic (23.90 ± 3.84 years and BMI 37.92 ± 4.85 kg/m2) and 28 age-matched healthy lean (24.56 ± 3.50 years and BMI of 22.10 ± 1.72 kg/m2) men were included in this study. The primary outcome measures were serum basal ANGPTL3 and ANGPTL3-area under the curve (AUC) levels. The percentage of body fat was measured by dual-energy X-ray absorptiometry and biochemical, hormonal and insulin resistance indices were determined. Basal ANGPTL3 and ANGPTL3-AUC levels were significantly elevated (p < 0.05) in young obese subjects compared with lean subjects and were positively and significantly associated with different anthropometric measurements. Fasting ANGPTL3 serum levels were positively correlated with fasting insulin, leptin, Leptin/Adiponectin index and triglyceride-glucose index. Moreover, ANGPTL3-AUC was negatively correlated with Matsuda index. In this regard, chronically high ANGPTL3 levels in young obese subjects might favor triglyceride-rich lipoprotein clearance to replenish triglyceride stores by white adipose tissue rather than oxidative tissues.
Sigma-Aldrich
Human ANGPTL3 / Angiopoietin-related Protein 3 ELISA Kit