MilliporeSigma
  • Assessment of the potential allergenicity and toxicity of Pichia proteins in a novel leghemoglobin preparation.
Assessment of the potential allergenicity and toxicity of Pichia proteins in a novel leghemoglobin preparation.
Regulatory toxicology and pharmacology : RTP (2020-11-11)
Teresa F Reyes, Yiming Chen, Rachel Z Fraser, Teresa Chan, Xin Li
ABSTRACT
The production of soy leghemoglobin C2 (LegH) by Pichia pastoris (syn. K. phaffii) was developed by Impossible Foods to serve as a sustainable source of flavor and aroma in plant-based meats. The potential allergenicity and toxicity of a LegH from a new production process was analyzed using bioinformatics, proteomics and a pepsin digestion assay on leghemoglobin, and residual host proteins. LegH in the new preparation had the same proteoform as in the previous preparations as well as in soy root nodule extracts. Results of seven Pichia proteins, each representing ≥1% of the total protein content, showed no significant sequence matches to any known allergens with the exception of one, which matched the highly conserved wheat GAPDH, whose protein homolog is found in fungi and humans. Based on the data, it is unlikely that there is any risk of cross reactivity between LegH Prep and GAPDH. Pichia protein sequences showed very good alignment to homologous proteins from many common yeasts including Saccharomyces sp. In addition, LegH and Pichia proteins were all rapidly digested in a pepsin digest assay. In conclusion, LegH Prep from this P. pastoris production process is unlikely to pose a risk of food allergenicity.
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Pricing and availability is not currently available.