MilliporeSigma
  • Comparing the Degradation Potential of Copper(II), Iron(II), Iron(III) Oxides, and Their Composite Nanoparticles in a Heterogeneous Photo-Fenton System.
Comparing the Degradation Potential of Copper(II), Iron(II), Iron(III) Oxides, and Their Composite Nanoparticles in a Heterogeneous Photo-Fenton System.
Nanomaterials (Basel, Switzerland) (2021-01-21)
Asfandyar Khan, Zsolt Valicsek, Ottó Horváth
ABSTRACT
Heterogeneous photo-Fenton systems offer efficient solutions for the treatment of wastewaters in the textile industry. This study investigated the fabrication and structural characterization of novel peculiar-shaped CuIIO, FeIII 2O3, and FeIIO nanoparticles (NPs) compared to the properties of the iron(II)-doped copper ferrite CuII 0.4FeII 0.6FeIII 2O4. The photocatalytic efficiencies of these NPs and the composite of the simple oxides (CuIIO/FeIIO/FeIII 2O3) regarding the degradation of methylene blue (MB) and rhodamine B (RhB) as model dyes were also determined. The catalysts were synthesized via simple co-precipitation and calcination technique. X-ray diffractometry (XRD), scanning electron microscopy (SEM), and diffuse reflectance spectroscopy (DRS) were utilized for structural characterization. The structure of CuIIO was bead-like connected into threads, FeIII 2O3 was rod-like, while FeIIO pallet-like, with average crystallite sizes of 18.9, 36.9, and 37.1 nm, respectively. The highest degradation efficiency was achieved by CuIIO for RhB and by CuII 0.4FeII 0.6FeIII 2O4 for MB. The CuIIO/FeIIO/FeIII 2O3 composite proved to be the second-best catalyst in both cases, with excellent reusability. Hence, these NPs can be successfully applied as heterogeneous photo-Fenton catalysts for the removal of hazardous pollutants. Moreover, the simple metal oxides and the iron(II)-doped copper ferrite displayed a sufficient antibacterial activity against Gram-negative Vibrio fischeri.
Product Description
Methylthioninium chloride, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.