MilliporeSigma
  • Low-dose carboplatin reprograms tumor immune microenvironment through STING signaling pathway and synergizes with PD-1 inhibitors in lung cancer.
Low-dose carboplatin reprograms tumor immune microenvironment through STING signaling pathway and synergizes with PD-1 inhibitors in lung cancer.
Cancer letters (2020-12-06)
Li Zhou, Qiuli Xu, Litang Huang, Jiajia Jin, Xueying Zuo, Qun Zhang, Liang Ye, Suhua Zhu, Ping Zhan, Jianan Ren, Tangfeng Lv, Yong Song
ABSTRACT
Although the combination of chemotherapy and immunotherapy is a hot topic in lung cancer, little is understood regarding the possible mechanisms behind their synergy. Moreover, safety is a major concern for clinicians while performing chemotherapy. Therefore, it is important to determine the appropriate dose and period of chemotherapy for combining it with immunotherapy, and investigate the underlying synergistic mechanism. Here, we showed that carboplatin can induce DNA damage and activate the canonical STING/TBK1/IRF3 pathway and non-canonical STING-NF-κB signaling complex. Further, low-dose carboplatin changed the "cold" tumor into a "hot" tumor via the signaling hub STING, augmenting CD8+ T-cell infiltration, increasing PD-L1 expression, and hence potentiating the anti-tumor effect of PD-1 inhibitors; importantly, there were no adverse effects. Furthermore, knocking down STING in tumor cells effectively reversed PD-L1 upregulation and STING pathway activation, and reduced the anti-tumor effect of low-dose carboplatin and carboplatin-PD-1 inhibitor combination. Our findings collectively reported a previously unexplored role of low-dose carboplatin targeting in the STING pathway and provided an economical, useful and safe option for improving the efficacy of PD-1 inhibitors in lung cancer.
Sigma-Aldrich
MISSION® esiRNA, targeting human TMEM173
Pricing and availability is not currently available.