MilliporeSigma
  • Surveillance as Determinant of Long-Term Survival in Non-Transplanted Hepatocellular Carcinoma Patients.
Surveillance as Determinant of Long-Term Survival in Non-Transplanted Hepatocellular Carcinoma Patients.
Cancers (2021-03-07)
Filippo Pelizzaro, Alessandro Vitale, Anna Sartori, Andrea Vieno, Barbara Penzo, Francesco Paolo Russo, Anna Chiara Frigo, Edoardo G Giannini, Manuela Piccinnu, Gian Ludovico Rapaccini, Maria Di Marco, Eugenio Caturelli, Marco Zoli, Rodolfo Sacco, Ciro Celsa, Fabio Marra, Andrea Mega, Maria Guarino, Antonio Gasbarrini, Gianluca Svegliati-Baroni, Francesco Giuseppe Foschi, Andrea Olivani, Alberto Masotto, Pietro Coccoli, Giovanni Raimondo, Francesco Azzaroli, Gianpaolo Vidili, Maurizia Rossana Brunetto, Franco Trevisani, Fabio Farinati, On Behalf Of Ita Li Ca Study Group
ABSTRACT
We aimed at assessing the impact of surveillance on long-term survival in HCC patients. From the ITA.LI.CA database, we selected 1028 cases with long (≥5 years, LS group) and 2721 controls with short-term survival (<5 years, SS group). The association between surveillance and LS was adjusted for confounders by multivariable logistic regression analysis. Survival of surveilled patients was presented both as observed and corrected for the lead-time bias, and the comparison of survival between surveillance and no surveillance groups was also performed after balancing the baseline characteristics with inverse probability weights (IPW). LS patients were more frequently diagnosed under surveillance (p < 0.0001), and had more favorable baseline characteristics. Surveillance was an independent predictor of LS (OR = 1.413, 95% CI 1.195-1.671; p < 0.0001). The observed and the lead-time corrected survival of surveilled patients were significantly longer compared to the survival of not surveilled patients (p < 0.0001 and p = 0.0008, respectively). In IPW adjusted populations, no survival differences were demonstrated between the two groups (p = 0.30). Surveillance, increasing early-stage diagnosis and applicability of curative treatments, is a fundamental determinant of long-term survival in HCC patients. A wide implementation of surveillance programs should be pursued in order to improve HCC patients' prognosis.
Sigma-Aldrich
N,O-Bis(trimethylsilyl)carbamate, ≥98.0% (T)